siRNA Details
| Gene Symbol: Nme1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138548.2 | NP_612557.1 | 2 | 218 | ||
| GenBank mRNA | D13374.1 | - | 2 | 219 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nme1| Gene Symbol: Nme1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138548.2 | NP_612557.1 | 2 | 218 | ||
| GenBank mRNA | D13374.1 | - | 2 | 219 | ||