siRNA Details
Gene Symbol: Ehd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139324.1 | NP_647540.1 | 3, 4 | 543 | ||
GenBank mRNA | AY094093.1 | - | 3, 4 | 543 | ||
BC083175.1 | - | 3, 4 | 525 | 7105816 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ehd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139324.1 | NP_647540.1 | 3, 4 | 543 | ||
GenBank mRNA | AY094093.1 | - | 3, 4 | 543 | ||
BC083175.1 | - | 3, 4 | 525 | 7105816 |