siRNA Details
Gene Symbol: Pigl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138901.2 | NP_620256.1 | 1 | 166 | ||
XM_008767767.1 | XP_008765989.1 | 1 | 162 | |||
GenBank mRNA | BC074020.1 | - | 1 | 166 | 7104008 | |
D88364.1 | - | 1 | 547 |
