siRNA Details
| Gene Symbol: Stip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138911.3 | NP_620266.1 | 12 | 1386 | ||
| GenBank mRNA | BC061529.1 | - | 12 | 1387 | 6887497 | |
| FQ220758.1 | - | 9 | 908 | |||
| FQ227512.1 | - | 3 | 137 | |||
| Y15068.1 | - | 11 | 1321 | |||

Stip1
Genomic Map