siRNA Details
Gene Symbol: Ppp1r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138823.2 | NP_620178.1 | 738 | |||
GenBank mRNA | BC092194.1 | - | 738 | 7313160 | ||
S79213.1 | - | 753 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ppp1r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138823.2 | NP_620178.1 | 738 | |||
GenBank mRNA | BC092194.1 | - | 738 | 7313160 | ||
S79213.1 | - | 753 |