siRNA Details
Gene Symbol: Prokr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138977.1 | NP_620433.1 | 2 | 1047 | ||
XM_006236821.2 | XP_006236883.1 | 3 | 1342 | |||
GenBank mRNA | AB158419.1 | - | 2 | 1047 | ||
AB158420.1 | - | 2 | 1047 | |||
AY089974.1 | - | 2 | 1047 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
