siRNA Details
| Gene Symbol: Adh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF508795.1 | - | 5 | 535 | ||
| BC062403.1 | - | 5 | 507 | 5621394 | ||
| CK480694.1 | - | 5 | 500 | |||
| FQ209590.1 | - | 5 | 518 | |||
| FQ218571.1 | - | 5 | 520 | |||
| FQ220674.1 | - | 5 | 518 | |||
| M15327.1 | - | 5 | 491 | |||
| X72792.1 | - | 3 | 321 | |||

Adh1
Genomic Map