siRNA Details
| Gene Symbol: Adra2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138506.1 | NP_612515.1 | 1 | 1422 | ||
| GenBank mRNA | M58316.1 | - | 1 | 1422 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adra2c| Gene Symbol: Adra2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138506.1 | NP_612515.1 | 1 | 1422 | ||
| GenBank mRNA | M58316.1 | - | 1 | 1422 | ||
Set Membership: |
|