siRNA Details
| Gene Symbol: Alpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022665.3 | NP_073156.3 | 9 | 1182 | ||
| XM_008767194.1 | XP_008765416.1 | 10 | 1217 | |||
| GenBank mRNA | X17611.1 | - | 9 | 1181 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alpi| Gene Symbol: Alpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022665.3 | NP_073156.3 | 9 | 1182 | ||
| XM_008767194.1 | XP_008765416.1 | 10 | 1217 | |||
| GenBank mRNA | X17611.1 | - | 9 | 1181 | ||