siRNA Details
| Gene Symbol: C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172222.2 | NP_757376.2 | 10 | 1422 | ||
| GenBank mRNA | AY149994.1 | - | 10 | 1332 | ||
| BC070923.1 | - | 10 | 1422 | 7096131 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C2| Gene Symbol: C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172222.2 | NP_757376.2 | 10 | 1422 | ||
| GenBank mRNA | AY149994.1 | - | 10 | 1332 | ||
| BC070923.1 | - | 10 | 1422 | 7096131 | ||