siRNA Details
| Gene Symbol: C3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016994.2 | NP_058690.2 | 26 | 3356 | ||
| GenBank mRNA | CO796620.1 | - | 3 | 223 | ||
| X52477.1 | - | 26 | 3356 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C3| Gene Symbol: C3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016994.2 | NP_058690.2 | 26 | 3356 | ||
| GenBank mRNA | CO796620.1 | - | 3 | 223 | ||
| X52477.1 | - | 26 | 3356 | |||