siRNA Details
| Gene Symbol: Camk2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012519.2 | NP_036651.1 | 19 | 1704 | ||
| XM_006233284.2 | XP_006233346.1 | 18 | 1863 | |||
| XM_006233285.2 | XP_006233347.1 | 18 | 1854 | |||
| XM_008761452.1 | XP_008759674.1 | 19 | 1923 | |||
| GenBank mRNA | BC107562.1 | - | 1464 | 7939424 | ||
| J05072.1 | - | 1704 | ||||
| L13406.1 | - | 5 | 441 | |||
| L13407.1 | - | 6 | 474 | |||
| L13408.1 | - | 6 | 501 | |||

Camk2d
Genomic Map