siRNA Details
| Gene Symbol: Ckb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012529.3 | NP_036661.3 | 649 | |||
| GenBank mRNA | BC065307.1 | - | 631 | 5598839 | ||
| BC070955.1 | - | 636 | 7104205 | |||
| BC087656.1 | - | 648 | ||||
| BC099814.1 | - | 633 | 7929525 | |||
| BC127477.1 | - | 639 | 8367180 | |||
| FQ216091.1 | - | 3 | 368 | |||
| M14400.1 | - | 661 | ||||
| M57664.1 | - | 4 | 512 | |||

Ckb
Genomic Map