siRNA Details
| Gene Symbol: Crygc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081660.1 | NP_001075129.1 | 3 | 297 | ||
| NM_033483.1 | NP_264077.1 | 2 | 280 | |||
| GenBank mRNA | AI717452.1 | - | 1 | 334 | ||
| BF523642.1 | - | 2 | 286 | |||
| BG371711.1 | - | 1 | 343 | |||

Crygc
Genomic Map