siRNA Details
| Gene Symbol: Egr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012551.2 | NP_036683.1 | 2 | 1180 | ||
| GenBank mRNA | AY551092.1 | - | 2 | 1180 | ||
| M18416.1 | - | 1532 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Egr1| Gene Symbol: Egr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012551.2 | NP_036683.1 | 2 | 1180 | ||
| GenBank mRNA | AY551092.1 | - | 2 | 1180 | ||
| M18416.1 | - | 1532 | ||||