siRNA Details
| Gene Symbol: Gck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270849.1 | NP_001257778.1 | 9 | 1115 | ||
| NM_001270850.1 | NP_001257779.1 | 8 | 893 | |||
| NM_012565.2 | NP_036697.1 | 8 | 1016 | |||
| XM_006251179.2 | XP_006251241.1 | 8 | 1861 | |||
| GenBank mRNA | J04218.1 | - | 8 | 975 | ||
| KJ026953.1 | - | 8 | 908 | |||
| M25807.1 | - | 8 | 893 | |||
| X53588.1 | - | 8 | 999 | |||
| X53589.1 | - | 9 | 1115 | |||

Gck
Genomic Map