siRNA Details
| Gene Symbol: Gck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270849.1 | NP_001257778.1 | 4, 5 | 567 | ||
| NM_012565.2 | NP_036697.1 | 3, 4 | 468 | |||
| XM_006251179.2 | XP_006251241.1 | 3, 4 | 1313 | |||
| GenBank mRNA | J04218.1 | - | 3, 4 | 427 | ||
| KJ026953.1 | - | 3, 4 | 360 | |||
| M58759.1 | - | 3, 4 | 850 | |||
| X53588.1 | - | 3, 4 | 451 | |||
| X53589.1 | - | 4, 5 | 567 | |||

Gck
Genomic Map