siRNA Details
| Gene Symbol: Gfi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012566.1 | NP_036698.1 | 4 | 831 | ||
| XM_008769917.1 | XP_008768139.1 | 4 | 1072 | |||
| XM_008769918.1 | XP_008768140.1 | 4 | 907 | |||
| XM_008769919.1 | XP_008768141.1 | 4 | 1789 | |||
| XM_008769920.1 | XP_008768142.1 | 4 | 1789 | |||
| GenBank mRNA | L06986.1 | - | 4 | 831 | ||

Gfi1
Genomic Map