siRNA Details
| Gene Symbol: Gnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030987.2 | NP_112249.2 | 10 | 952 | ||
| XM_006239517.2 | XP_006239579.1 | 10 | 1104 | |||
| XM_006239518.2 | XP_006239580.1 | 10 | 1104 | |||
| XM_006239519.2 | XP_006239581.1 | 10 | 1104 | |||
| XM_006239520.2 | XP_006239582.1 | 10 | 1104 | |||
| XM_008764335.1 | XP_008762557.1 | 9 | 987 | |||
| GenBank mRNA | AY552805.1 | - | 8 | 804 | ||
| BC078809.1 | - | 10 | 952 | 7123318 | ||
| DQ120483.1 | - | 5 | 557 | |||
| DQ120484.1 | - | 5 | 557 | |||
| U34958.1 | - | 8 | 804 | |||
| U88324.1 | - | 8 | 841 | |||

Gnb1
Genomic Map