siRNA Details
| Gene Symbol: Gspt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003978.1 | NP_001003978.1 | 12 | 1659 | ||
| GenBank mRNA | BC079092.1 | - | 12 | 1659 | 7123818 | |
| Gene Symbol: LOC100911685 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750763.3 | XP_003750811.1 | 2082 | |||
| XM_006245740.2 | XP_006245802.1 | 2085 | ||||

Gspt1
Genomic Map