siRNA Details
| Gene Symbol: Gstm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177426.1 | NP_803175.1 | 64 | |||
| GenBank mRNA | BC158576.1 | - | 2 | 87 | ||
| FJ179402.1 | - | 64 | ||||
| FQ214985.1 | - | 2 | 97 | |||
| FQ218547.1 | - | 2 | 130 | |||
| FQ218596.1 | - | 2 | 97 | |||
| FQ219549.1 | - | 2 | 99 | |||
| FQ219560.1 | - | 2 | 97 | |||

Gstm2
Genomic Map