siRNA Details
Gene Symbol: Irf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012591.1 | NP_036723.1 | 2 | 235 | ||
GenBank mRNA | BC076382.1 | - | 2 | 148 | 7099391 | |
M34253.1 | - | 2 | 235 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Irf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012591.1 | NP_036723.1 | 2 | 235 | ||
GenBank mRNA | BC076382.1 | - | 2 | 148 | 7099391 | |
M34253.1 | - | 2 | 235 |