siRNA Details
Gene Symbol: Mog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022668.2 | NP_073159.2 | 6, 8 | 802 | ||
XM_008772687.1 | XP_008770909.1 | 6, 7 | 1559 | |||
GenBank mRNA | BC087717.1 | - | 6, 8 | 802 | 7317044 | |
L21995.1 | - | 5, 6 | 633 | |||
M99485.1 | - | 6, 8 | 802 |
