siRNA Details
| Gene Symbol: Neu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031522.3 | NP_113710.2 | 5 | 1152 | ||
| GenBank mRNA | AB035722.1 | - | 5 | 1010 | ||
| BC100639.1 | - | 5 | 1027 | 7929913 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_010893, Y11412, U93702, AK145680, AK148229, and others.

Neu1
Genomic Map