siRNA Details
| Gene Symbol: Ctsq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139262.1 | NP_640355.1 | 5, 6 | 592 | ||
| GenBank mRNA | AF187323.1 | - | 5, 6 | 592 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctsq| Gene Symbol: Ctsq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139262.1 | NP_640355.1 | 5, 6 | 592 | ||
| GenBank mRNA | AF187323.1 | - | 5, 6 | 592 | ||