siRNA Details
Gene Symbol: Tas2r126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139335.1 | NP_647551.1 | 1 | 773 | ||
GenBank mRNA | AF240768.1 | - | 1 | 773 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tas2r126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139335.1 | NP_647551.1 | 1 | 773 | ||
GenBank mRNA | AF240768.1 | - | 1 | 773 |