siRNA Details
Gene Symbol: Zfp382 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144749.1 | NP_653350.1 | 1389 | |||
GenBank mRNA | BC127483.1 | - | 802 | 8365988 | ||
U56732.1 | - | 1389 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp382 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144749.1 | NP_653350.1 | 1389 | |||
GenBank mRNA | BC127483.1 | - | 802 | 8365988 | ||
U56732.1 | - | 1389 |