siRNA Details
Gene Symbol: Aspg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144750.2 | NP_653351.1 | 12 | 1447 | ||
GenBank mRNA | AB009372.1 | - | 12 | 1448 | ||
BC098655.1 | - | 12 | 1417 | 7373402 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Aspg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144750.2 | NP_653351.1 | 12 | 1447 | ||
GenBank mRNA | AB009372.1 | - | 12 | 1448 | ||
BC098655.1 | - | 12 | 1417 | 7373402 |