siRNA Details
| Gene Symbol: Serbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145086.1 | NP_659554.1 | 1, 2 | 369 | ||
| XM_006236616.1 | XP_006236678.1 | 1, 2 | 417 | |||
| XM_006236617.1 | XP_006236679.1 | 1, 2 | 417 | |||
| XM_006236618.1 | XP_006236680.1 | 1, 2 | 417 | |||
| GenBank mRNA | AF388527.1 | - | 1, 2 | 375 | ||
| BC079337.1 | - | 1, 2 | 369 | 7106027 | ||
| FQ213573.1 | - | 1, 2 | 412 | |||
| FQ214321.1 | - | 1, 2 | 368 | |||
| FQ223736.1 | - | 1, 2 | 390 | |||
| FQ225988.1 | - | 1, 2 | 84 | |||
| FQ226374.1 | - | 1, 2 | 390 | |||
| FQ226420.1 | - | 1, 2 | 84 | |||
| FQ227636.1 | - | 1, 2 | 390 | |||
| FQ228026.1 | - | 1, 2 | 380 | |||
| FQ230135.1 | - | 1, 2 | 396 | |||
| FQ230192.1 | - | 1, 2 | 380 | |||
| FQ230382.1 | - | 1, 2 | 83 | |||
| FQ231329.1 | - | 1, 2 | 84 | |||
| FQ231840.1 | - | 1, 2 | 84 | |||
| FQ232288.1 | - | 1, 2 | 390 | |||
| FQ232894.1 | - | 1, 2 | 391 | |||
| FQ233179.1 | - | 1, 2 | 390 | |||
| FQ233457.1 | - | 1, 2 | 396 | |||
| FQ233487.1 | - | 1, 2 | 84 | |||
| FQ233911.1 | - | 1, 2 | 390 | |||
| FQ234284.1 | - | 1, 2 | 364 | |||
| FQ234534.1 | - | 1, 2 | 391 | |||
| FQ235054.1 | - | 1, 2 | 84 | |||

Serbp1
Genomic Map