siRNA Details
| Gene Symbol: Arfgap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145090.3 | NP_659558.1 | 7 | 793 | ||
| GenBank mRNA | DQ167810.1 | - | 5 | 510 | ||
| DQ167811.1 | - | 5 | 510 | |||
| U35776.1 | - | 7 | 793 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arfgap1| Gene Symbol: Arfgap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145090.3 | NP_659558.1 | 7 | 793 | ||
| GenBank mRNA | DQ167810.1 | - | 5 | 510 | ||
| DQ167811.1 | - | 5 | 510 | |||
| U35776.1 | - | 7 | 793 | |||