siRNA Details
| Gene Symbol: Nrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145098.2 | NP_659566.1 | 15 | 2680 | ||
| XM_006255825.2 | XP_006255887.1 | 15 | 2873 | |||
| XM_006255826.2 | XP_006255888.1 | 14 | 2637 | |||
| XM_006255827.2 | XP_006255889.1 | 14 | 2658 | |||
| XM_008772598.1 | XP_008770820.1 | 14 | 2658 | |||
| GenBank mRNA | AF016296.1 | - | 14 | 2284 | ||
| AF018957.1 | - | 14 | 2137 | |||
| BC085689.1 | - | 15 | 2680 | 7109697 | ||

Nrp1
Genomic Map