siRNA Details
| Gene Symbol: Allc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013037.1 | NP_001013055.1 | 8 | 857 | ||
| XM_006239944.2 | XP_006240006.1 | 8 | 931 | |||
| GenBank mRNA | AF480062.1 | - | 5 | 511 | ||
| BC078971.1 | - | 8 | 857 | 7111813 | ||

Allc
Genomic Map