siRNA Details
| Gene Symbol: Slc25a25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145677.1 | NP_663710.1 | 1 | 149 | ||
| XM_006233935.2 | XP_006233997.1 | 1 | 1486 | |||
| GenBank mRNA | AY043169.1 | - | 1 | 149 | ||
| FQ211394.1 | - | 1 | 165 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a25| Gene Symbol: Slc25a25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145677.1 | NP_663710.1 | 1 | 149 | ||
| XM_006233935.2 | XP_006233997.1 | 1 | 1486 | |||
| GenBank mRNA | AY043169.1 | - | 1 | 149 | ||
| FQ211394.1 | - | 1 | 165 | |||