siRNA Details
| Gene Symbol: Prps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012634.1 | NP_036766.1 | 6 | 1080 | ||
| XM_006256829.2 | XP_006256891.1 | 6 | 852 | |||
| GenBank mRNA | M17259.1 | - | 6 | 821 | ||
| M29393.1 | - | 6 | 821 | |||
| X16555.1 | - | 6 | 1080 | |||

Prps2
Genomic Map