siRNA Details
| Gene Symbol: Rara | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031528.2 | NP_113716.2 | 5 | 986 | ||
| XM_008767945.1 | XP_008766167.1 | 5 | 786 | |||
| GenBank mRNA | AJ002940.1 | - | 5 | 710 | ||
| AJ002941.1 | - | 5 | 726 | |||
| BC099830.1 | - | 5 | 986 | 7933868 | ||
| U15211.1 | - | 5 | 992 | |||

Rara
Genomic Map