siRNA Details
| Gene Symbol: Rho | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033441.1 | NP_254276.1 | 3 | 704 | ||
| GenBank mRNA | Z46957.1 | - | 3 | 704 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_145383, BC013125, AK044333, BC031766, AK044412.
Set Membership: |
|

Rho
Genomic Map