siRNA Details
Gene Symbol: Slc2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138827.1 | NP_620182.1 | 5 | 850 | ||
GenBank mRNA | BC061873.1 | - | 5 | 778 | 5598829 | |
M13979.1 | - | 5 | 850 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138827.1 | NP_620182.1 | 5 | 850 | ||
GenBank mRNA | BC061873.1 | - | 5 | 778 | 5598829 | |
M13979.1 | - | 5 | 850 |