siRNA Details
| Gene Symbol: Slc4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012651.2 | NP_036783.2 | 16 | 2176 | ||
| XM_008767946.1 | XP_008766168.1 | 16 | 2179 | |||
| GenBank mRNA | AY030082.1 | - | 13 | 1798 | ||
| BC085748.1 | - | 13 | 2187 | 7126960 | ||

Slc4a1
Genomic Map