siRNA Details
| Gene Symbol: Stat5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017064.1 | NP_058760.1 | 13, 14 | 1741 | ||
| GenBank mRNA | U24175.1 | - | 13, 14 | 1741 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stat5a| Gene Symbol: Stat5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017064.1 | NP_058760.1 | 13, 14 | 1741 | ||
| GenBank mRNA | U24175.1 | - | 13, 14 | 1741 | ||