siRNA Details
| Gene Symbol: Rbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012733.4 | NP_036865.1 | 2 | 278 | ||
| GenBank mRNA | FQ209770.1 | - | 2 | 292 | ||
| FQ209927.1 | - | 2 | 262 | |||
| FQ210835.1 | - | 2 | 293 | |||
| FQ218090.1 | - | 2 | 292 | |||
| M16459.1 | - | 2 | 278 | |||
| M19257.1 | - | 2 | 208 | |||

Rbp1
Genomic Map