siRNA Details
| Gene Symbol: Crabp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105716.1 | NP_001099186.1 | 4 | 490 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crabp1| Gene Symbol: Crabp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105716.1 | NP_001099186.1 | 4 | 490 | ||