siRNA Details
| Gene Symbol: Tnfrsf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019135.1 | NP_062008.1 | 13 | 1352 | ||
| GenBank mRNA | D42117.1 | - | 13 | 1352 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnfrsf8| Gene Symbol: Tnfrsf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019135.1 | NP_062008.1 | 13 | 1352 | ||
| GenBank mRNA | D42117.1 | - | 13 | 1352 | ||