siRNA Details
| Gene Symbol: Cyp2e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031543.1 | NP_113731.1 | 6 | 890 | ||
| XM_006230520.1 | XP_006230582.1 | 7 | 1073 | |||
| GenBank mRNA | AF061442.1 | - | 6 | 878 | ||
| BC081774.1 | - | 6 | 892 | 7110096 | ||
| FQ212595.1 | - | 3 | 256 | |||
| FQ218528.1 | - | 4 | 529 | |||
| FQ219179.1 | - | 3 | 256 | |||
| J02627.1 | - | 6 | 890 | |||

Cyp2e1
Genomic Map