siRNA Details
| Gene Symbol: Klrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012745.2 | NP_036877.1 | 6 | 715 | ||
| GenBank mRNA | AF009133.1 | - | 6 | 472 | ||
| BC086393.1 | - | 6 | 715 | 7303354 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klrd1| Gene Symbol: Klrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012745.2 | NP_036877.1 | 6 | 715 | ||
| GenBank mRNA | AF009133.1 | - | 6 | 472 | ||
| BC086393.1 | - | 6 | 715 | 7303354 | ||