siRNA Details
Gene Symbol: Gadd45a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024127.2 | NP_077041.2 | 3, 4 | 538 | ||
GenBank mRNA | AB102787.1 | - | 3, 4 | 388 | ||
AB102788.1 | - | 3, 4 | 388 | |||
BC081795.1 | - | 3, 4 | 538 | 7110642 | ||
L32591.1 | - | 3, 4 | 406 |
