siRNA Details
| Gene Symbol: Ccnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC059113.1 | - | 2 | 253 | 6922392 | |
| FN800625.1 | - | 2 | 251 | |||
| L11995.1 | - | 2 | 245 | |||
| X60768.1 | - | 2 | 183 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccnb1| Gene Symbol: Ccnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC059113.1 | - | 2 | 253 | 6922392 | |
| FN800625.1 | - | 2 | 251 | |||
| L11995.1 | - | 2 | 245 | |||
| X60768.1 | - | 2 | 183 | |||