siRNA Details
| Gene Symbol: Chrm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080773.1 | NP_542951.1 | 1 | 434 | ||
| XM_006230974.2 | XP_006231036.1 | 2 | 1127 | |||
| XM_008760215.1 | XP_008758437.1 | 2 | 946 | |||
More Information
Set Membership: |
|

Chrm1
Genomic Map