siRNA Details
| Gene Symbol: Cnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012809.2 | NP_036941.1 | 2 | 726 | ||
| XM_006247260.2 | XP_006247322.1 | 2 | 746 | |||
| XM_006247261.2 | XP_006247323.1 | 2 | 750 | |||
| XM_008767948.1 | XP_008766170.1 | 2 | 660 | |||
| GenBank mRNA | BC098066.1 | - | 2 | 726 | 7458826 | |
| L16532.1 | - | 2 | 725 | |||

Cnp
Genomic Map