siRNA Details
| Gene Symbol: Miox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145771.2 | NP_665714.2 | 8, 9 | 653 | ||
| XM_008765674.1 | XP_008763896.1 | 8, 9 | 837 | |||
| GenBank mRNA | AF197128.1 | - | 8, 9 | 647 | ||
| AF230096.1 | - | 7, 8 | 655 | |||
| AY738259.1 | - | 7, 8 | 647 | |||
| BC078840.1 | - | 8, 9 | 653 | 7110224 | ||
| FQ219187.1 | - | 7, 8 | 642 | |||

Miox
Genomic Map