siRNA Details
| Gene Symbol: Miox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145771.2 | NP_665714.2 | 2, 3 | 92 | ||
| XM_008765674.1 | XP_008763896.1 | 2, 3 | 276 | |||
| GenBank mRNA | AF197128.1 | - | 2, 3 | 86 | ||
| AY738259.1 | - | 1, 2 | 86 | |||
| BC078840.1 | - | 2, 3 | 92 | 7110224 | ||
| FQ219187.1 | - | 1, 2 | 81 | |||

Miox
Genomic Map